Tricholine citrate ∼65% in water

供应商: TCI

Synonyms: Choline Citrate Solution

C0330-25G C0330-500G
TCIAC0330-25GEA 0 CNY
TCIAC0330-25G TCIAC0330-500G
Tricholine citrate ∼65% in water
Tricholine citrate
MDL编号: MFCD00059985
CAS号: 546-63-4
EINECS: 208-907-5

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Concentration (Nonaqueous Titration) 60.0 to 66.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR