2-甲基-4-氯苯氧乙酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: MCPA, 4-氯-邻甲苯氧基乙酸

C0206-25G C0206-500G
TCIAC0206-25GEA 0 CNY
TCIAC0206-25G TCIAC0206-500G
2-甲基-4-氯苯氧乙酸 ≥98.0% (by titrimetric analysis)
2-甲基-4-氯苯氧乙酸
分子式: C₉H₉ClO₃
分子量: 200.62 g/mol
MDL编号: MFCD00004306
CAS号: 94-74-6
EINECS: 202-360-6
Merck Index: 14,05764

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 118.0 to 121.0 deg-C
Solubility in Methanol within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR