Chloranilic acid lanthanum(III) salt decahydrate ≥98.0% (通过滴定分析)

供应商: TCI

C0079-25G C0079-5G
TCIAC0079-25GEA 0 CNY
TCIAC0079-25G TCIAC0079-5G
Chloranilic acid lanthanum(III) salt decahydrate ≥98.0% (通过滴定分析)
Chloranilic acid lanthanum(III) salt decahydrate
分子式: C₆H₂Cl₂O₄
MDL编号: MFCD00135764
CAS号: 32607-23-1
EINECS: 251-127-5

现在订购

产品技术规格

Appearance Black powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Water 15.0 to 18.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR