2-Bromo-3-hexyl-5-iodothiophene ≥97.0% (by GC) stabilised
供应商: TCI
Stabilization:
Stabilised with copper chip
B3865-1G
B3865-5G
TCIAB3865-1GEA
0
CNY
TCIAB3865-1G
TCIAB3865-5G
2-Bromo-3-hexyl-5-iodothiophene ≥97.0% (by GC) stabilised
2-Bromo-3-hexyl-5-iodothiophene
分子式:
C₁₀H₁₄BrIS 分子量: 373.09 g/mol 沸点: 121°C 存储条件: Freezer |
MDL编号:
MFCD15144651 CAS号: 160096-76-4 |
产品技术规格
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...