3-溴-2,4,6-三甲基苯胺 ≥98.0% (by GC, titration analysis)
供应商: TCI
Warning
B3849-5G B3849-25G
TCIAB3849-5GEA 0 CNY
TCIAB3849-5G TCIAB3849-25G
3-溴-2,4,6-三甲基苯胺 ≥98.0% (by GC, titration analysis)
3-溴-2,4,6-三甲基苯胺
分子式: C₉H₁₂BrN
分子量: 214.11 g/mol
沸点: 173°C
存储条件: Refrigerator
MDL编号: MFCD00015476
CAS号: 82842-52-2
现在订购
产品技术规格
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 36.0 to 40.0 deg-C