Boc-Chg-OH ≥98.0% (by HPLC)
供应商: TCI
Synonyms:
Boc-L-α-环己基甘氨酸
B3839-1G
B3839-5G
TCIAB3839-1GEA
0
CNY
TCIAB3839-1G
TCIAB3839-5G
Boc-Chg-OH ≥98.0% (by HPLC)
Boc-Chg-OH
分子式:
C₁₃H₂₃NO₄ 分子量: 257.3 g/mol 溶点: 83°C |
MDL编号:
MFCD00133638 CAS号: 109183-71-3 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Specific rotation [a]20/D | +9.0 to +12.0 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...