3-丁烯酸叔丁酯 ≥95.0% (by GC)

供应商: TCI
Warning

Synonyms: 乙烯基乙酸叔丁酯

B3811-5G B3811-25G
TCIAB3811-5GEA 0 CNY
TCIAB3811-5G TCIAB3811-25G
3-丁烯酸叔丁酯 ≥95.0% (by GC)
3-丁烯酸叔丁酯
分子式: C₈H₁₄O₂
分子量: 142.20 g/mol
闪点: 31°C
MDL编号: MFCD06411714
CAS号: 14036-55-6
UN号: 3272
ADR: 3 III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR