Butein ≥98.0% (by HPLC)
供应商: TCI
Synonyms:
2',3,4,4'-Tetrahydroxychalcone, (E)-1-(2,4-Dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)-2-propen-1-one
B3803-1G
B3803-100MG
TCIAB3803-1GEA
0
CNY
TCIAB3803-1G
TCIAB3803-100MG
Butein ≥98.0% (by HPLC)
Butein
分子式:
C₁₅H₁₂O₅ 分子量: 272.26 g/mol 溶点: 215°C 存储条件: Freezer |
MDL编号:
MFCD00017300 CAS号: 487-52-5 EINECS: 207-659-5 |
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...