3-Bromocatechol ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
3-Bromobenzene-1,2-diol, 1-Bromo-2,3-dihydroxybenzene
B3710-1G
B3710-5G
TCIAB3710-1GEA
0
CNY
TCIAB3710-1G
TCIAB3710-5G
3-Bromocatechol ≥98.0% (by GC)
3-Bromocatechol
分子式:
C₆H₅BrO₂ 分子量: 189.01 g/mol 沸点: 120°C |
MDL编号:
MFCD00869768 CAS号: 14381-51-2 |
产品技术规格
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...