5-tert-Butyl-1-methyl-L-glutamate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
供应商: TCI
Synonyms:
L-Glutamic Acid 5-tert-Butyl 1-Methyl Ester Hydrochloride, H-Glu(OtBu)-OMe·HCl, H-Glu(OtBu)-OMe.HCl
B3670-1G
B3670-5G
TCIAB3670-1GEA
0
CNY
TCIAB3670-1G
TCIAB3670-5G
5-tert-Butyl-1-methyl-L-glutamate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
5-tert-Butyl-1-methyl-L-glutamate hydrochloride
分子式:
C₁₀H₁₉NO₄ 分子量: 253.72 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00153438 CAS号: 6234-01-1 EINECS: 228-348-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Specific rotation [a]20/D | +24.0 to +28.0 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...