N-Fmoc-D-glutamic acid 5-tert-butyl ester ≥98.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamate, 5-tert-Butyl N-Fmoc-D-glutamate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic Acid 5-tert-Butyl Ester, Fmoc-D-Glu(OtBu)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid
B3669-1G
B3669-5G
TCIAB3669-1GEA
0
CNY
TCIAB3669-1G
TCIAB3669-5G
N-Fmoc-D-glutamic acid 5-tert-butyl ester ≥98.0% (by HPLC, titration analysis)
N-Fmoc-D-glutamic acid 5-tert-butyl ester
分子式:
C₂₄H₂₇NO₆ 分子量: 425.48 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00077055 CAS号: 104091-08-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Melting point | 90.0 to 94.0 deg-C |
Specific rotation [a]20/D | +7.0 to +10.0 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...