N-Boc-2-哌啶甲酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
(±)-1-叔丁氧羰基-哌啶-2-羧酸, Boc-DL-Pip-OH, N-Boc-DL-2-哌啶甲酸
B3644-25G
B3644-5G
TCIAB3644-25GEA
0
CNY
TCIAB3644-25G
TCIAB3644-5G
N-Boc-2-哌啶甲酸 ≥98.0% (by GC, titration analysis)
N-Boc-2-哌啶甲酸
分子式:
C₁₁H₁₉NO₄ 分子量: 229.276 g/mol |
MDL编号:
MFCD01862877 CAS号: 98303-20-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 129.0 to 133.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...