2,2′-联吡啶-3,3′-二羧酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
2,2'-Binicotinic Acid, [2,2'-Bipyridine]-3,3'-dicarboxylic acid
B3622-1G
B3622-5G
TCIAB3622-1GEA
0
CNY
TCIAB3622-1G
TCIAB3622-5G
2,2′-联吡啶-3,3′-二羧酸 ≥98.0% (by GC, titration analysis)
2,2′-联吡啶-3,3′-二羧酸
分子式:
C₁₂H₈N₂O₄ 溶点: 261°C |
MDL编号:
MFCD00226068 CAS号: 4433-01-6 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...