5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt ≥98.0% (by HPLC, titration analysis) for biochemical research
供应商: TCI
Synonyms:
XGLUC Sodium Salt, 5-Bromo-4-chloro-3-indolyl-b-D-glucuronide sodium salt
B3621-10MG
B3621-100MG
TCIAB3621-10MGEA
0
CNY
TCIAB3621-10MG
TCIAB3621-100MG
5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt ≥98.0% (by HPLC, titration analysis) for biochemical research
5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide sodium salt
分子式:
C₁₄H₁₃BrClNO₇ 分子量: 444.59 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00135782 CAS号: 129541-41-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
beta-Glucuronidase detection test | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...