Boc-Phe(4-Cl)-OH ≥98.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
Boc-L-4-氯苯丙氨酸
B3614-1G
B3614-5G
TCIAB3614-1GEA
0
CNY
TCIAB3614-1G
TCIAB3614-5G
Boc-Phe(4-Cl)-OH ≥98.0% (by HPLC, titration analysis)
Boc-Phe(4-Cl)-OH
分子式:
C₁₄H₁₈ClNO₄ 分子量: 299.75 g/mol |
MDL编号:
MFCD00057808 CAS号: 68090-88-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 110.0 to 114.0 deg-C |
Specific rotation [a]20/D | +22.0 to +26.0 deg (C=1, EtOAc) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...