1,3-Bis(3-aminophenoxy)-5-chlorobenzene dihydrochloride ≥98.0% (by HPLC)

供应商: TCI

Synonyms: 3,3'-((5-Chloro-1,3-phenylene)bis(oxy))dianiline dihydrochloride

B3550-1G B3550-5G
TCIAB3550-1GEA 0 CNY
TCIAB3550-1G TCIAB3550-5G
1,3-Bis(3-aminophenoxy)-5-chlorobenzene dihydrochloride ≥98.0% (by HPLC)
1,3-Bis(3-aminophenoxy)-5-chlorobenzene dihydrochloride
分子式: C₁₈H₁₅ClN₂O₂
分子量: 399.70 g/mol
MDL编号: MFCD14708170
CAS号: 99503-73-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR