顺式-3-叔丁氧羰酰胺环己羧酸 ≥97.0% (by titrimetric analysis)

供应商: TCI

Synonyms: cis-3-(Boc-amino)cyclohexanecarboxylic Acid, cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, cis-3-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid

B3518-1G B3518-5G
TCIAB3518-1GEA 0 CNY
TCIAB3518-1G TCIAB3518-5G
顺式-3-叔丁氧羰酰胺环己羧酸 ≥97.0% (by titrimetric analysis)
顺式-3-叔丁氧羰酰胺环己羧酸
分子式: C₁₂H₂₁NO₄
分子量: 243.30 g/mol
MDL编号: MFCD03453252
CAS号: 222530-33-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 143.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR