1,1′-二茂铁二硼酸二(频哪醇)酯 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
1,1′-二茂铁二硼酸频哪醇酯
B3501-1G
B3501-5G
TCIAB3501-1GEA
0
CNY
TCIAB3501-1G
TCIAB3501-5G
1,1′-二茂铁二硼酸二(频哪醇)酯 ≥98.0% (by GC, titration analysis)
1,1′-二茂铁二硼酸二(频哪醇)酯
分子式:
C₂₂H₃₂B₂FeO₄ |
MDL编号:
MFCD06411336 CAS号: 737776-93-1 |
产品技术规格
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Chelometric Titration) | min. 98.0 % |
Melting point | 171.0 to 175.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...