4-tert-Butyldiphenyl sulphide ≥98.0% (通过GC)

供应商: TCI
Warning

B3484-100G B3484-25G
TCIAB3484-100GEA 0 CNY
TCIAB3484-100G TCIAB3484-25G
4-tert-Butyldiphenyl sulphide ≥98.0% (通过GC)
4-tert-Butyldiphenyl sulphide
分子式: C₁₆H₁₈S
沸点: 140°C
MDL编号: MFCD06200845
CAS号: 85609-03-6

现在订购

产品技术规格

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR