1,3-双(2,6-二异丙基苯基)咪唑-2-烯 ≥98.0% (通过HPLC,滴定分析)
供应商: TCI
|
Warning
|
B3465-1G
B3465-5G
TCIAB3465-1GEA
0
CNY
TCIAB3465-1G
TCIAB3465-5G
1,3-双(2,6-二异丙基苯基)咪唑-2-烯 ≥98.0% (通过HPLC,滴定分析)
1,3-双(2,6-二异丙基苯基)咪唑-2-烯
分子式:
C₂₇H₃₆N₂ 存储条件: Refrigerator |
MDL编号:
MFCD07781991 CAS号: 244187-81-3 |
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...