苄基硼酸频哪醇酯 ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-苄基-4,4,5,5-四甲基-(1,3,2)二氧杂戊硼烷

B3448-1G B3448-5G
TCIAB3448-1GEA 0 CNY
TCIAB3448-1G TCIAB3448-5G
苄基硼酸频哪醇酯 ≥97.0% (by GC)
苄基硼酸频哪醇酯
分子式: C₁₃H₁₉BO₂
分子量: 218.10 g/mol
MDL编号: MFCD05663841
CAS号: 87100-28-5

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR