1-Butyl-1-methylpiperidinium bromide ≥97.0% (by titrimetric analysis)

供应商: TCI

Synonyms: N-butyl-N-methyl-piperidinium bromide

B3424-25G B3424-5G
TCIAB3424-25GEA 0 CNY
TCIAB3424-25G TCIAB3424-5G
1-Butyl-1-methylpiperidinium bromide ≥97.0% (by titrimetric analysis)
1-Butyl-1-methylpiperidinium bromide
分子式: C₁₀H₂₂BrN
分子量: 236.20 g/mol
溶点: 230°C
MDL编号: MFCD11870112
CAS号: 94280-72-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR