[3S-(3α,4aβ,8aβ)]-N-(叔丁基)十氢-3-异喹啉甲酰胺 ≥97.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: [3S-(3α,4αβ,8αβ)]-N-(tert-Butyl)decahydroisoquinoline-3-carboxamide, (3S,4aS,8aS)-N-(tert-Butyl)decahydroisoquinoline-3-carboxamide

B3410-25G B3410-5G
TCIAB3410-25GEA 0 CNY
TCIAB3410-25G TCIAB3410-5G
[3S-(3α,4aβ,8aβ)]-N-(叔丁基)十氢-3-异喹啉甲酰胺 ≥97.0% (by GC, titration analysis)
[3S-(3α,4aβ,8aβ)]-N-(叔丁基)十氢-3-异喹啉甲酰胺
分子式: C₁₄H₂₆N₂O
分子量: 238.38 g/mol
MDL编号: MFCD01313226
CAS号: 136465-81-1
EINECS: 420-380-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 113.0 to 117.0 deg-C
Specific rotation [a]20/D -70.0 to -76.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR