(1E)-1-tert-Butoxy-1-(trimethylsilyloxy)propene ≥92.0% (by GC)
供应商: TCI
|
Warning
|
B3298-1G
B3298-5G
TCIAB3298-1GEA
0
CNY
TCIAB3298-1G
TCIAB3298-5G
(1E)-1-tert-Butoxy-1-(trimethylsilyloxy)propene ≥92.0% (by GC)
(1E)-1-tert-Butoxy-1-(trimethylsilyloxy)propene
分子式:
C₁₀H₂₂O₂Si 沸点: 53°C 存储条件: Refrigerator |
CAS号:
72658-10-7 UN号: 1993 ADR: 3,III |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 92.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...