Fmoc-Ser(tBu)-OH ≥98.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
Fmoc-O-叔丁基-L-丝氨酸
B3168-25G
B3168-5G
TCIAB3168-25GEA
0
CNY
TCIAB3168-25G
TCIAB3168-5G
Fmoc-Ser(tBu)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-Ser(tBu)-OH
分子式:
C₂₂H₂₅NO₅ 分子量: 383.44 g/mol |
MDL编号:
MFCD00037127 CAS号: 71989-33-8 EINECS: 276-260-6 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | +22.0 to +27.0 deg (C=1, EtOAc) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...