Nɑ-(tert-Butoxycarbonyl)-D-lysine ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: Nɑ-Boc-D-lysine, Boc-D-Lys-OH

B3083-1G B3083-5G
TCIAB3083-1GEA 0 CNY
TCIAB3083-1G TCIAB3083-5G
Nɑ-(tert-Butoxycarbonyl)-D-lysine ≥98.0% (by titrimetric analysis)
Nalpha-(tert-Butoxycarbonyl)-D-lysine
分子式: C₁₁H₂₂N₂O₄
分子量: 246.31 g/mol
MDL编号: MFCD00076956
CAS号: 106719-44-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -19.0 to -23.0 deg (C=2, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR