4-Bromo-N,N-3,5-tetramethylaniline ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-Bromo-5-(dimethylamino)-m-xylene, 4-Bromo-N,N,3,5-tetramethylaniline, 4-Bromo-N,N-dimethyl-3,5-xylidine
B3070-1G
B3070-5G
TCIAB3070-1GEA
0
CNY
TCIAB3070-1G
TCIAB3070-5G
4-Bromo-N,N-3,5-tetramethylaniline ≥98.0% (by GC)
4-Bromo-N,N-3,5-tetramethylaniline
分子式:
C₁₀H₁₄BrN 分子量: 228.13 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD11036245 CAS号: 14275-09-3 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...