2-甲基吡啶硼烷复合物 ≥85.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: 2-Picoline Borane, 2-Methylpyridine Borane, Borane - 2-Picoline Complex

B3018-25G B3018-5G
TCIAB3018-25GEA 0 CNY
TCIAB3018-25G TCIAB3018-5G
2-甲基吡啶硼烷复合物 ≥85.0% (by titrimetric analysis)
2-甲基吡啶硼烷复合物
分子式: C₆H₁₀BN
分子量: 106.96 g/mol
MDL编号: MFCD07784361
CAS号: 3999-38-0
UN号: 2813
ADR: 4.3 II

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (Potassium iodate Method) min. 85.0 %
Melting point 44.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR