Boc-D-Asn-OH ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: Boc-D-天冬酰胺

B2964-25G B2964-5G
TCIAB2964-25GEA 0 CNY
TCIAB2964-25G TCIAB2964-5G
Boc-D-Asn-OH ≥98.0% (by HPLC, titration analysis)
Boc-D-Asn-OH
分子式: C₉H₁₆N₂O₅
分子量: 232.24 g/mol
MDL编号: MFCD00065558
CAS号: 75647-01-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 99.0 ee%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +6.0 to +8.0 deg (C=1, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR