1,6-己二醇二丙烯酸酯 ≥85.0% (by GC) stabilised
供应商: TCI
|
Warning
|
Synonyms:
HDODA
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
B2936-25G
B2936-500G
TCIAB2936-25GEA
0
CNY
TCIAB2936-25G
TCIAB2936-500G
1,6-己二醇二丙烯酸酯 ≥85.0% (by GC) stabilised
1,6-己二醇二丙烯酸酯
分子式:
C₁₂H₁₈O₄ 分子量: 226.27 g/mol 沸点: 165°C |
MDL编号:
MFCD00008631 CAS号: 13048-33-4 EINECS: 235-921-9 UN号: 3082 ADR: 9 III |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 85.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...