Boc-5-Ava-OH ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
5-(Boc-氨基)戊酸, 5-(Boc-氨基)颉草酸
B2895-25G
B2895-5G
TCIAB2895-25GEA
0
CNY
TCIAB2895-25G
TCIAB2895-5G
Boc-5-Ava-OH ≥98.0% (by GC, titration analysis)
Boc-5-Ava-OH
分子式:
C₁₀H₁₉NO₄ 分子量: 217.27 g/mol 沸点: 168°C |
MDL编号:
MFCD00076903 CAS号: 27219-07-4 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...