苄氧乙酸 ≥97.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 2-(Benzyloxy)acetic acid

B2876-1G B2876-5G
TCIAB2876-1GEA 0 CNY
TCIAB2876-1G TCIAB2876-5G
苄氧乙酸 ≥97.0% (by GC, titration analysis)
苄氧乙酸
分子式: C₉H₁₀O₃
分子量: 166.18 g/mol
沸点: 155°C
MDL编号: MFCD00274211
CAS号: 30379-55-6

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR