Bis(trimethylsilyl)bromomethane ≥95.0% (by GC)

供应商: TCI
Danger

Synonyms: Bis(trimethylsilyl)methyl Bromide

B2874-1G B2874-5G
TCIAB2874-1GEA 0 CNY
TCIAB2874-1G TCIAB2874-5G
Bis(trimethylsilyl)bromomethane ≥95.0% (by GC)
Bis(trimethylsilyl)bromomethane
分子式: C₇H₁₉BrSi₂
分子量: 239.30 g/mol
存储条件: Refrigerator
CAS号: 29955-12-2
UN号: 1760
ADR: 8 III

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR