5-(Boc-氨基)-1-戊醇 ≥97.0% (by GC)

供应商: TCI

Synonyms: N-(5-羟基戊基)氨基甲酸叔丁酯

B2869-25G B2869-5G
TCIAB2869-25GEA 0 CNY
TCIAB2869-25G TCIAB2869-5G
5-(Boc-氨基)-1-戊醇 ≥97.0% (by GC)
5-(Boc-氨基)-1-戊醇
分子式: C₁₀H₂₁NO₃
分子量: 203.28 g/mol
MDL编号: MFCD01862955
CAS号: 75178-90-4

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR