Brassinazole ≥93.0% (by HPLC)

供应商: TCI
Warning

Synonyms: 1-[2-(4-Chlorophenyl)-1-(1-hydroxy-1-phenylethyl)ethyl]-1,2,4-triazole, 3-(4-Chlorophenyl)-1-methyl-1-phenyl-2-(1,2,4-triazol-1-yl)-1-propanol, 4-(4-Chlorophenyl)-2-phenyl-3-(1H-1,2,4-triazol-1-yl)butan-2-ol

B2829-10MG B2829-100MG
TCIAB2829-10MGEA 0 CNY
TCIAB2829-10MG TCIAB2829-100MG
Brassinazole ≥93.0% (by HPLC)
Brassinazole
分子式: C₁₈H₁₈ClN₃O
MDL编号: MFCD08276319
CAS号: 224047-41-0

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 93.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR