1,2-Bis(2,3,4,5,6-pentabromophenyl)ethane ≥96.0% (by titrimetric analysis)
供应商: TCI
Synonyms:
2,2',3,3',4,4',5,5',6,6'-Decabromobibenzyl, 1,2-Bis(perbromophenyl)ethane
B2795-25G
B2795-500G
TCIAB2795-25GEA
0
CNY
TCIAB2795-25G
TCIAB2795-500G
1,2-Bis(2,3,4,5,6-pentabromophenyl)ethane ≥96.0% (by titrimetric analysis)
1,2-Bis(2,3,4,5,6-pentabromophenyl)ethane
分子式:
C₁₄H₄Br₁₀ 分子量: 971.23 g/mol |
MDL编号:
MFCD06407713 CAS号: 84852-53-9 EINECS: 284-366-9 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Argentmetric Titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...