Bis(4-cyanophenyl)ether ≥98.0% (通过GC)

供应商: TCI
Danger

B2790-25G B2790-5G
TCIAB2790-25GEA 0 CNY
TCIAB2790-25G TCIAB2790-5G
Bis(4-cyanophenyl)ether ≥98.0% (通过GC)
Bis(4-cyanophenyl)ether
分子式: C₁₄H₈N₂O
沸点: 220°C
CAS号: 6508-04-9
EINECS: 229-399-1
UN号: 3439
ADR: 6.1,III

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 182.0 to 186.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR