2,4-双(α,α-二甲基苯甲基)苯酚 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2,4-Bis(2-phenylpropan-2-yl)phenol, 2,4-Di-ɑ-cumylphenol
B2772-25G
B2772-500G
TCIAB2772-25GEA
0
CNY
TCIAB2772-25G
TCIAB2772-500G
2,4-双(α,α-二甲基苯甲基)苯酚 ≥97.0% (by GC)
2,4-双(α,α-二甲基苯甲基)苯酚
分子式:
C₂₄H₂₆O 分子量: 330.47 g/mol 沸点: 221°C |
MDL编号:
MFCD00191242 CAS号: 2772-45-4 EINECS: 220-466-0 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 64.0 to 67.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...