1-双(4-氟苯基)甲基哌嗪 ≥97.0% (by GC)

供应商: TCI
Danger

Synonyms: 1-(4,4′-二氟二苯甲基)哌嗪

B2764-25G B2764-5G
TCIAB2764-25GEA 0 CNY
TCIAB2764-25G TCIAB2764-5G
1-双(4-氟苯基)甲基哌嗪 ≥97.0% (by GC)
1-双(4-氟苯基)甲基哌嗪
分子式: C₁₇H₁₈F₂N₂
分子量: 288.34 g/mol
沸点: 130°C
MDL编号: MFCD00038660
CAS号: 27469-60-9
EINECS: 248-476-0
UN号: 2811
ADR: 6.1 III

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 91.0 to 95.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR