1,4-Bis(2-benzoxazolyl)naphthalene ≥98.0% (by HPLC, total nitrogen)

供应商: TCI

Synonyms: 1,4-Bis(benzo[d]oxazol-2-yl)naphthalene

B2728-25G B2728-500G
TCIAB2728-25GEA 0 CNY
TCIAB2728-25G TCIAB2728-500G
1,4-Bis(2-benzoxazolyl)naphthalene ≥98.0% (by HPLC, total nitrogen)
1,4-Bis(2-benzoxazolyl)naphthalene
分子式: C₂₄H₁₄N₂O₂
MDL编号: MFCD00357168
CAS号: 5089-22-5
EINECS: 225-803-5

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 211.0 to 215.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR