Bis(2-butoxyethyl)adipate ≥97.0% (by GC)
供应商: TCI
Synonyms:
Di(2-butoxyethyl) Adipate, Adipic Acid Bis(2-butoxyethyl) Ester, Adipic Acid Di(2-butoxyethyl) Ester
B2716-25G
B2716-500G
TCIAB2716-25GEA
0
CNY
TCIAB2716-25G
TCIAB2716-500G
Bis(2-butoxyethyl)adipate ≥97.0% (by GC)
Bis(2-butoxyethyl)adipate
分子式:
C₁₈H₃₄O₆ 分子量: 346.46 g/mol 沸点: 217°C 溶点: –34°C 闪点: 199°C |
CAS号:
141-18-4 EINECS: 205-466-0 |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...