Benzyl-2-chloroethyl ether ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-(Benzyloxy)ethyl Chloride, ((2-Chloroethoxy)methyl)benzene, (2-Chloroethoxymethyl)benzene

B2712-25G B2712-5G
TCIAB2712-25GEA 0 CNY
TCIAB2712-25G TCIAB2712-5G
Benzyl-2-chloroethyl ether ≥98.0% (by GC)
Benzyl-2-chloroethyl ether
分子式: C₉H₁₁ClO
分子量: 170.64 g/mol
MDL编号: MFCD00018964
CAS号: 17229-17-3

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR