N-(4-Bromophenyl)phthalimide ≥98.0% (by HPLC, total nitrogen)

供应商: TCI
Warning

Synonyms: 2-(4-Bromophenyl)isoindoline-1,3-dione

B2707-25G B2707-5G
TCIAB2707-25GEA 0 CNY
TCIAB2707-25G TCIAB2707-5G
N-(4-Bromophenyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
N-(4-Bromophenyl)phthalimide
分子式: C₁₄H₈BrNO₂
分子量: 302.13 g/mol
MDL编号: MFCD00047301
CAS号: 40101-31-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 203.0 to 209.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR