9,9-Bis(4-amino-3-methylphenyl)fluorene ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 4,4'-(9H-Fluorene-9,9-diyl)bis(2-methylaniline), 2,2'-Dimethyl-4,4'-(9-fluorenylidene)dianiline, 4,4'-(9-Fluorenylidene)di-o-toluidine

B2693-25G B2693-5G
TCIAB2693-25GEA 0 CNY
TCIAB2693-25G TCIAB2693-5G
9,9-Bis(4-amino-3-methylphenyl)fluorene ≥98.0% (by titrimetric analysis)
9,9-Bis(4-amino-3-methylphenyl)fluorene
分子式: C₂₇H₂₄N₂
MDL编号: MFCD04038168
CAS号: 107934-60-1

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 230.0 to 236.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR