3,5-Bis(tert-butylthio)-1-chlorobenzene ≥98.0% (by GC)

供应商: TCI

Synonyms: 1-Chloro-3,5-bis(tert-butylthio)benzene

B2659-1G B2659-5G
TCIAB2659-1GEA 0 CNY
TCIAB2659-1G TCIAB2659-5G
3,5-Bis(tert-butylthio)-1-chlorobenzene ≥98.0% (by GC)
3,5-Bis(tert-butylthio)-1-chlorobenzene
分子式: C₁₄H₂₁ClS₂
分子量: 288.89 g/mol
CAS号: 260968-02-3

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Solubility in Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR