4-氟苯基醚 ≥98.0% (by GC)

供应商: TCI

Synonyms: 4,4'-Oxybis(fluorobenzene), 4,4'-Difluorodiphenyl Ether

B2648-1G B2648-5G
TCIAB2648-1GEA 0 CNY
TCIAB2648-1G TCIAB2648-5G
4-氟苯基醚 ≥98.0% (by GC)
4-氟苯基醚
分子式: C₁₂H₈F₂O
沸点: 240°C
MDL编号: MFCD00013552
CAS号: 330-93-8
EINECS: 206-358-6

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR