1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene ≥98.0% (by HPLC)
供应商: TCI
B2629-1G
B2629-100MG
TCIAB2629-1GEA
0
CNY
TCIAB2629-1G
TCIAB2629-100MG
1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene ≥98.0% (by HPLC)
1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene
分子式:
C₂₉H₂₂F₆S₂ |
CAS号:
172612-67-8 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 129.0 to 133.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...