2,2′-联嘧啶 ≥95.0% (通过GC)

供应商: TCI
Warning

B2496-1G B2496-200MG
TCIAB2496-1GEA 0 CNY
TCIAB2496-1G TCIAB2496-200MG
2,2′-联嘧啶 ≥95.0% (通过GC)
2,2′-联嘧啶
分子式: C₈H₆N₄
溶点: 113°C
MDL编号: MFCD00014600
CAS号: 34671-83-5
EINECS: 252-137-2

现在订购

产品技术规格

Appearance White to Green to Brown powder to crystal
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR