双(2,4,6-三甲基吡啶)碘(I)六氟磷酸盐 ≥98.0% (by HPLC)

供应商: TCI
Warning

B2359-1G B2359-5G
TCIAB2359-1GEA 0 CNY
TCIAB2359-1G TCIAB2359-5G
双(2,4,6-三甲基吡啶)碘(I)六氟磷酸盐 ≥98.0% (by HPLC)
双(2,4,6-三甲基吡啶)碘(I)六氟磷酸盐
分子式: C₁₆H₂₂F₆IN₂P
存储条件: Refrigerator
CAS号: 113119-46-3

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR