4-Bromocrotonic acid ≥98.0% (通过GC,滴定分析)

供应商: TCI
Danger

B2298-1G B2298-5G
TCIAB2298-1GEA 0 CNY
TCIAB2298-1G TCIAB2298-5G
4-Bromocrotonic acid ≥98.0% (通过GC,滴定分析)
4-Bromocrotonic acid
分子式: C₄H₅BrO₂
溶点: 74°C
存储条件: Refrigerator
MDL编号: MFCD00082701
CAS号: 13991-36-1
UN号: 3261
ADR: 8,III

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 73.0 to 77.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR